RIE156328820 Rieke Metals, Inc.

8-(2,5-difluorophenyl)-8-oxooctanoic acid

  • MDL number MFCD01311649
  • Cas Number : 898766-40-0
  • Molecular Weight : 270.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7067F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7067F
Availability details for
Quantity
  • Availability
  • Add to Cart