RIE156328821 Rieke Metals, Inc.

4-(2,6-Difluorophenyl)-4-oxobutyric acid

  • MDL number MFCD01320111
  • Cas Number : 478162-62-8
  • Molecular Weight : 214.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7068B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7068B
Availability details for
Quantity
  • Availability
  • Add to Cart