RIE156328822 Rieke Metals, Inc.

5-(2,6-difluorophenyl)-5-oxovaleric acid

  • MDL number MFCD01320112
  • Cas Number : 898766-43-3
  • Molecular Weight : 228.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7068C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7068C
Availability details for
Quantity
  • Availability
  • Add to Cart