RIE156328825 Rieke Metals, Inc.

8-(2,6-difluorophenyl)-8-oxooctanoic acid

  • MDL number MFCD01311668
  • Cas Number : 898766-52-4
  • Molecular Weight : 270.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7068F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7068F
Availability details for
Quantity
  • Availability
  • Add to Cart