RIE156328826 Rieke Metals, Inc.

5-(2-Fluorophenyl)-5-oxovaleric acid

  • MDL number MFCD01320116
  • Cas Number : 199664-70-5
  • Molecular Weight : 210.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7069C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7069C
Availability details for
Quantity
  • Availability
  • Add to Cart