RIE156328827 Rieke Metals, Inc.

6-(2-fluorophenyl)-6-oxohexanoic acid

  • MDL number MFCD01320117
  • Cas Number : 56721-59-6
  • Molecular Weight : 224.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7069D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7069D
Availability details for
Quantity
  • Availability
  • Add to Cart