RIE156328831 Rieke Metals, Inc.

7-(4-biphenyl)-7-oxoheptanoic acid

  • MDL number MFCD01320129
  • Cas Number : 362670-19-7
  • Molecular Weight : 296.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7072E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7072E
Availability details for
Quantity
  • Availability
  • Add to Cart