RIE156328832 Rieke Metals, Inc.

2,2-Dimethyl-4-(3-methoxyphenyl)-4-oxobutyric acid

  • MDL number MFCD01320130
  • Cas Number : 898766-61-5
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7073

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7073
Availability details for
Quantity
  • Availability
  • Add to Cart