RIE156328834 Rieke Metals, Inc.

2,2-dimethyl-4-(2-methoxyphenyl)-4-oxobutyric acid

  • MDL number MFCD01320132
  • Cas Number : 898766-64-8
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7075

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7075
Availability details for
Quantity
  • Availability
  • Add to Cart