RIE156328835 Rieke Metals, Inc.

4-(3,4-Difluorophenyl)-2,2-dimethyl-4-oxobutyric acid

  • MDL number MFCD01320133
  • Cas Number : 898766-66-0
  • Molecular Weight : 242.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7076

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7076
Availability details for
Quantity
  • Availability
  • Add to Cart