RIE156328837 Rieke Metals, Inc.

2,2-dimethyl-4-(3-fluorophenyl)-4-oxobutyric acid

  • MDL number MFCD01320135
  • Cas Number : 898766-70-6
  • Molecular Weight : 224.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7078

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7078
Availability details for
Quantity
  • Availability
  • Add to Cart