RIE156328838 Rieke Metals, Inc.

2,2-dimethyl-4-oxo-4-(3,4,5-trifluorophenyl)butryic acid

  • MDL number MFCD07700143
  • Cas Number : 898766-72-8
  • Molecular Weight : 260.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7079

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7079
Availability details for
Quantity
  • Availability
  • Add to Cart