RIE156328839 Rieke Metals, Inc.

2,2-Dimethyl-4-(2-naphthyl)-4-oxobutyric acid

  • MDL number MFCD01320137
  • Cas Number : 854648-79-6
  • Molecular Weight : 256.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7080

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7080
Availability details for
Quantity
  • Availability
  • Add to Cart