RIE156328847 Rieke Metals, Inc.

6-Cyclobutyl-6-oxohexanoic acid

  • MDL number MFCD01320149
  • Cas Number : 898766-81-9
  • Molecular Weight : 184.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7084D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7084D
Availability details for
Quantity
  • Availability
  • Add to Cart