RIE156328859 Rieke Metals, Inc.

5-(4-Chlorophenyl)-5-oxovaleric acid

  • MDL number MFCD01320019
  • Cas Number : 36978-49-1
  • Molecular Weight : 226.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7004C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7004C
Availability details for
Quantity
  • Availability
  • Add to Cart