RIE156328860 Rieke Metals, Inc.

2'-Methoxy-2,2,2-trifluoroacetophenone

  • MDL number MFCD00461894
  • Cas Number : 26944-43-4
  • Molecular Weight : 204.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6503-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6503-1
Availability details for
Quantity
  • Availability
  • Add to Cart