RIE156328862 Rieke Metals, Inc.

4'-Methoxy-2,2,2-trifluoroacetophenone

  • MDL number MFCD00018061
  • Cas Number : 711-38-6
  • Molecular Weight : 204.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6503-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6503-3
Availability details for
Quantity
  • Availability
  • Add to Cart