RIE156328865 Rieke Metals, Inc.

5-chloro-1-(2-methoxyphenyl)-1-oxopentane

  • MDL number MFCD07700086
  • Cas Number : 43228-96-2
  • Molecular Weight : 226.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6142D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6142D
Availability details for
Quantity
  • Availability
  • Add to Cart