RIE156328882 Rieke Metals, Inc.

8-(4-Methoxyphenyl)-8-oxooctanoic acid

  • MDL number MFCD01311626
  • Cas Number : 22811-92-3
  • Molecular Weight : 264.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7011F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7011F
Availability details for
Quantity
  • Availability
  • Add to Cart