RIE156328891 Rieke Metals, Inc.

4'-t-Butyl-2,2,2-trifluoroacetophenone

  • MDL number MFCD01319979
  • Cas Number : 73471-97-3
  • Molecular Weight : 230.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6510

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6510
Availability details for
Quantity
  • Availability
  • Add to Cart