RIE156328892 Rieke Metals, Inc.

4'-ethyl-2,2,2-trifluoroacetophenone

  • MDL number MFCD01319980
  • Cas Number : 73471-96-2
  • Molecular Weight : 202.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6511A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6511A
Availability details for
Quantity
  • Availability
  • Add to Cart