RIE156328898 Rieke Metals, Inc.

2,2-Dimethyl-4-oxo-4-phenylbutyric acid

  • MDL number MFCD01320038
  • Cas Number : 15116-34-4
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7022

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7022
Availability details for
Quantity
  • Availability
  • Add to Cart