RIE156328899 Rieke Metals, Inc.

4-(2-Methylphenyl)-4-oxobutyric acid

  • MDL number MFCD00029848
  • Cas Number : 6939-36-2
  • Molecular Weight : 192.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7023B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7023B
Availability details for
Quantity
  • Availability
  • Add to Cart