RIE156328900 Rieke Metals, Inc.

5-(2-methylphenyl)-5-oxovaleric acid

  • MDL number MFCD01320039
  • Cas Number : 36978-50-4
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7023C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7023C
Availability details for
Quantity
  • Availability
  • Add to Cart