RIE156328901 Rieke Metals, Inc.

1-Chloro-3-(3,4-dimethoxyphenyl)-3-oxopropane

  • MDL number MFCD00510412
  • Cas Number : 4693-38-3
  • Molecular Weight : 228.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6153B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6153B
Availability details for
Quantity
  • Availability
  • Add to Cart