RIE156328902 Rieke Metals, Inc.

2',6'-Dimethyl-2,2,2-trifluoroacetophenone

  • MDL number MFCD01319986
  • Cas Number : 121456-63-1
  • Molecular Weight : 202.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6512-4

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6512-4
Availability details for
Quantity
  • Availability
  • Add to Cart