RIE156328915 Rieke Metals, Inc.

4-(3,5-Dimethylphenyl)-4-oxobutyric acid

  • MDL number MFCD01320052
  • Cas Number : 36440-58-1
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7036B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7036B
Availability details for
Quantity
  • Availability
  • Add to Cart