RIE156328920 Rieke Metals, Inc.

4-(3,5-Dichlorophenyl)-4-oxobutyric acid

  • MDL number MFCD01320057
  • Cas Number : 66740-88-3
  • Molecular Weight : 247.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7038B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7038B
Availability details for
Quantity
  • Availability
  • Add to Cart