RIE156328922 Rieke Metals, Inc.

3-(1,3-Dioxolan-2-yl)benzophenone

  • MDL number MFCD02260438
  • Cas Number : 85366-46-7
  • Molecular Weight : 254.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6128-00-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6128-00-01
Availability details for
Quantity
  • Availability
  • Add to Cart