RIE156328923 Rieke Metals, Inc.

4-(2-methoxyphenyl)-4-oxobutyronitrile

  • MDL number MFCD07700096
  • Cas Number : 113522-19-3
  • Molecular Weight : 189.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6145B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6145B
Availability details for
Quantity
  • Availability
  • Add to Cart