RIE156328927 Rieke Metals, Inc.

4'-methyl-2,2,2,3'-tetrafluoroacetophenone

  • MDL number MFCD01320003
  • Cas Number : 845823-06-5
  • Molecular Weight : 206.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6525

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6525
Availability details for
Quantity
  • Availability
  • Add to Cart