RIE156328928 Rieke Metals, Inc.

3'-methyl-2,2,2,4'-tetrafluoroacetophenone

  • MDL number MFCD01320004
  • Cas Number : 845823-10-1
  • Molecular Weight : 206.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6526

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6526
Availability details for
Quantity
  • Availability
  • Add to Cart