RIE156328931 Rieke Metals, Inc.

5-(3-Fluorophenyl)-5-oxovaleric acid

  • MDL number MFCD01320063
  • Cas Number : 845790-38-7
  • Molecular Weight : 210.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7042C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7042C
Availability details for
Quantity
  • Availability
  • Add to Cart