RIE156328932 Rieke Metals, Inc.

1-naphthyl trifluoromethyl ketone

  • MDL number MFCD01320007
  • Cas Number : 6500-37-4
  • Molecular Weight : 224.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6529-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6529-1
Availability details for
Quantity
  • Availability
  • Add to Cart