RIE156328936 Rieke Metals, Inc.

5-(3-chlorophenyl)-5-oxovaleric acid

  • MDL number MFCD01320065
  • Cas Number : 75381-46-3
  • Molecular Weight : 226.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7044C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7044C
Availability details for
Quantity
  • Availability
  • Add to Cart