RIE156328937 Rieke Metals, Inc.

5-(4-Methoxyphenyl)-5-oxovaleronitrile

  • MDL number MFCD00448777
  • Cas Number : 26823-02-9
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6147C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6147C
Availability details for
Quantity
  • Availability
  • Add to Cart