RIE156328938 Rieke Metals, Inc.

4-Chloro-1-(3-methoxyphenyl)-1-oxobutane

  • MDL number MFCD07700090
  • Cas Number : 258882-48-3
  • Molecular Weight : 212.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6143C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6143C
Availability details for
Quantity
  • Availability
  • Add to Cart