RIE156328939 Rieke Metals, Inc.

5-Chloro-1-(3-methoxyphenyl)-1-oxopentane

  • MDL number MFCD07700091
  • Cas Number : 258882-49-4
  • Molecular Weight : 226.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6143D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6143D
Availability details for
Quantity
  • Availability
  • Add to Cart