RIE156328942 Rieke Metals, Inc.

2'-phenyl-2,2,2-trifluoroacetophenone

  • MDL number MFCD01320013
  • Cas Number : 302912-29-4
  • Molecular Weight : 250.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6533-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6533-1
Availability details for
Quantity
  • Availability
  • Add to Cart