RIE156328944 Rieke Metals, Inc.

4'-Phenoxy-2,2,2-trifluoroacetophenone

  • MDL number MFCD00042392
  • Cas Number : 70783-32-3
  • Molecular Weight : 266.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6534

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6534
Availability details for
Quantity
  • Availability
  • Add to Cart