RIE156328946 Rieke Metals, Inc.

5-(3,5-Difluorophenyl)-5-oxovaleric acid

  • MDL number MFCD01320075
  • Cas Number : 845790-50-3
  • Molecular Weight : 228.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7048C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7048C
Availability details for
Quantity
  • Availability
  • Add to Cart