RIE156328951 Rieke Metals, Inc.

4'-Chloro-3'-methyl-2,2,2-trifluoroacetophenone

  • MDL number MFCD02260828
  • Cas Number : 286017-71-8
  • Molecular Weight : 222.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6538

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6538
Availability details for
Quantity
  • Availability
  • Add to Cart