RIE156328952 Rieke Metals, Inc.

2-chloro-5-trifluoroacetylthiophene

  • MDL number MFCD02260829
  • Cas Number : 771-83-5
  • Molecular Weight : 214.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6539

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6539
Availability details for
Quantity
  • Availability
  • Add to Cart