RIE156328953 Rieke Metals, Inc.

Cyclohexyltrifluoroacetone

  • MDL number MFCD02260830
  • Cas Number : 457-49-8
  • Molecular Weight : 194.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6540

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6540
Availability details for
Quantity
  • Availability
  • Add to Cart