RIE156328954 Rieke Metals, Inc.

4-(4-Fluoro-3-methylphenyl)-4-oxobutyric acid

  • MDL number MFCD07700141
  • Cas Number : 349-22-4
  • Molecular Weight : 210.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7054B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7054B
Availability details for
Quantity
  • Availability
  • Add to Cart