RIE156328955 Rieke Metals, Inc.

5-(4-fluoro-3-methylphenyl)-5-oxovaleric acid

  • MDL number MFCD07700142
  • Cas Number : 898765-95-2
  • Molecular Weight : 224.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7054C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7054C
Availability details for
Quantity
  • Availability
  • Add to Cart