RIE156328958 Rieke Metals, Inc.

4-Oxo-4-(3-phenoxyphenyl)butyric acid

  • MDL number MFCD00963144
  • Cas Number : 54364-84-0
  • Molecular Weight : 270.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7056B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7056B
Availability details for
Quantity
  • Availability
  • Add to Cart