RIE156328960 Rieke Metals, Inc.

4'-chloro-2'-methyl-2,2,2-trifluoroacetophenone

  • MDL number MFCD06201807
  • Cas Number : 845823-13-4
  • Molecular Weight : 222.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6541

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6541
Availability details for
Quantity
  • Availability
  • Add to Cart