RIE156328961 Rieke Metals, Inc.

7-oxo-7-(3-phenoxyphenyl)heptanoic acid

  • MDL number MFCD02261019
  • Cas Number : 871127-76-3
  • Molecular Weight : 312.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7056E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7056E
Availability details for
Quantity
  • Availability
  • Add to Cart