RIE156328964 Rieke Metals, Inc.

5-oxo-5-(4-thiomethylphenyl)valeric acid

  • MDL number MFCD01320086
  • Cas Number : 845790-49-0
  • Molecular Weight : 238.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7057C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7057C
Availability details for
Quantity
  • Availability
  • Add to Cart